Learn More
GE Healthcare Whatman™ Sterile Mixed Cellulose Ester Membranes, ME 25 Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  GE Healthcare 10406800
 View more versions of this product
Catalog No. 09-800-951
Add to cart
Description
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Yes
Black
Mixed cellulose ester
0.45μm
3.1mm
130°C
47mm
Mixed Cellulose Ester Membrane
White
Without interleaving paper and pad
ME25
Medium
Sterile
4.3 to 5.0mg/cm2
Membrane butler