Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Whatman™ Syringe-Type Membrane Holder Available on GSA/VA Contract for Federal Government customers only.
Stainless steel membrane holder for use with a standard syringe
$181.00
Specifications
Material Stainless Steel
View More Specs
Products 2
Catalog Number Mfr. No. Material Diameter (Metric) Inner Price Quantity & Availability  
Catalog Number Mfr. No. Material Diameter (Metric) Inner Price Quantity & Availability  
09-924-549 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
1980002
Stainless Steel (SS) 25 mm Pack for $365.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
09-908-1
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
1980001
Stainless Steel 13mm Each for $181.00
Add to cart
Description
Description
  • Designed for the quick and easy clarification, sterilization, and removal of particulates from small volume samples (typically for HPLC applications)
  • holders contain PTFE gaskets and O-rings and allow the membrane to be autoclaved in place without the filter sticking to the holder.
  • Luer lock fittings connect to a standard syringe and offer convenience and ease of use for clarification, sterilization, and removal of particulates from small volumes of liquid
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
    Specifications
    Specifications
    Stainless Steel