Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Whatman™ TurboBlotter Transfer System, Small Kits Available on GSA/VA Contract for Federal Government customers only.
Downward blotting device turns the conventional Northern/Southern transfer stack upside down for rapid, high-resolution transfer of DNA and RNA
$290.00 - $323.00
Specifications
For Use With (Application) For DNA/RNA transfer
View More Specs
Includes: Includes device plus five sets of transfer materials for gel size indicated in ordering table. Each set consists of one sheet of Nytran™ SPC 0.45µm Nylon membrane, one wick of 3MM Chr blotting paper and a blotter stack composed of eight sheets of 3MM Chr blotting paper and 20 sheets of GB004 blotting paper. Refills, available separately, contain five sets of transfer materials.
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Type Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Type Price Quantity & Availability  
09-301-176 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10416300
10 x 15 cm Gel TurboBlotter™ Transfer System Nytran™ SPC 0.45μm TurboBlotter™ Kit
Each for $290.00
Add to cart
09-301-178
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10416304
11 x 14 cm Gel TurboBlotter™ Transfer System Nytran™ SPC 0.45μm TurboBlotter™ Kit
Each for $323.00
Add to cart
Description
Description
  • Takes advantage of gravity — no heavy weights are required on the top of the transfer stack, eliminating the messy set-up of standard upward capillary transfers
  • Has a smaller footprint than most homemade devices and is stackable — up to five units can be stacked during transfers
  • Allows for alkaline buffer transfers in one hour and neutral (SSC) transfers in three hours
  • Very easy to set up and works without power or vacuum source
  • Requires less buffer and blotting paper than conventional transfers
  • Reusable
  • Model is 12 x 16cm, for gels from 7 x 8cm to 11 x 14cm
Order Info
Refill packs containing additional membranes and papers required for further transfer are available separately (Cat. Nos. 09-301-177, -179, -181, -183, -185 and -189).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
For DNA/RNA transfer