Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Whatman™ TurboBlotter Transfer System, Small Kits Available on GSA/VA Contract for Federal Government customers only.

Downward blotting device turns the conventional Northern/Southern transfer stack upside down for rapid, high-resolution transfer of DNA and RNA

Manufacturer:  GE Healthcare 10416304

 View more versions of this product

Catalog No. 09-301-178


Add to cart

Includes: Includes device plus five sets of transfer materials for gel size indicated in ordering table. Each set consists of one sheet of Nytran™ SPC 0.45µm Nylon membrane, one wick of 3MM Chr blotting paper and a blotter stack composed of eight sheets of 3MM Chr blotting paper and 20 sheets of GB004 blotting paper. Refills, available separately, contain five sets of transfer materials.

Description

Description

  • Takes advantage of gravity — no heavy weights are required on the top of the transfer stack, eliminating the messy set-up of standard upward capillary transfers
  • Has a smaller footprint than most homemade devices and is stackable — up to five units can be stacked during transfers
  • Allows for alkaline buffer transfers in one hour and neutral (SSC) transfers in three hours
  • Very easy to set up and works without power or vacuum source
  • Requires less buffer and blotting paper than conventional transfers
  • Reusable

  • Model is 12 x 16cm, for gels from 7 x 8cm to 11 x 14cm

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Order Info

Refill packs containing additional membranes and papers required for further transfer are available separately (Cat. Nos. 09-301-177, -179, -181, -183, -185 and -189).

Specifications

Specifications

Nytran™ SPC 0.45μm TurboBlotter™ kit
4.3 in.
11cm
12 x 16cm Small transfer device, 5 sets of transfer materials (set consists of one sheet of Nytran™ SPC 0.45μm Nylon membrane, one wick of 3mm chromatography blotting Paper and a blotter stack composed of eight sheets o
5.5 in.
14cm
11 x 14cm Gel TurboBlotter™ transfer system
For DNA/RNA transfer
Documents
Promotions

Promotions

  • GE Healthcare Life Sciences , Fisherbrand
    Get 5% off your purchase of $500 to $999.99 in GE Healthcare™ Life Sciences products with promo code GESAVE5.
    : GESAVE5

    09-30-2018

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit