Learn More
Cytiva Whatman™ Type WCN Cellulose Nitrate Membranes
Recommended for majority of routine applications, this membrane is manufactured under strictly controlled conditions. Whatman™ Type WCN Cellulose Nitrate Membranes narrow pore size distribution for improved surface capture and analysis.
$185.80 - $715.00
Specifications
Autoclavable Autoclavable
Color White
Material Cellulose Nitrate
View More Specs
Products 24
Catalog Number Mfr. No. Pore Size Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Pore Size Diameter (Metric) Price Quantity & Availability  
09-924-582
View Documents
Cytiva
7190002
1.0 μm 25 mm
Pack of 100 for $248.00
Add to Cart
09-905-15
View Documents
Cytiva
7184002
0.45 μm 25 mm
Pack of 100 for $232.00
Add to Cart
09-924-579
View Documents
Cytiva
7188002
0.8 μm 25 mm
Pack of 100 for $185.80
Add to Cart
09-905-2
View Documents
Cytiva
7181004
0.1 μm 47 mm
Pack of 100 for $715.00
Add to Cart
09-905-45
View Documents
Cytiva
7190004
1.0 μm 47 mm
Pack of 100 for $477.00
Add to Cart
09-905-17
View Documents
Cytiva
7184004
0.45 μm 47 mm
Pack of 100 for $376.00
Add to Cart
09-924-583
View Documents
Cytiva
7191014
1.2 μm 142 mm
Pack of 25 for $392.00
Add to Cart
09-905-39
View Documents
Cytiva
7188004
0.8 μm 47 mm
Pack of 100 for $431.00
Add to Cart
09-905-56
View Documents
Cytiva
7195004
5 μm 47 mm
Pack of 100 for $477.00
Add to Cart
09-905-21
View Documents
Cytiva
7184014
0.45 μm 142 mm
Pack of 25 for $515.00
Add to Cart
09-905-50
View Documents
Cytiva
7193004
3.0 μm 47 mm
Pack of 100 for $457.00
Add to Cart
09-927-897
View Documents
Cytiva
7186004
0.65 μm 47 mm
Pack of 100 for $454.00
Add to Cart
09-924-580
View Documents
Cytiva
7188003
0.8 μm 37 mm
Pack of 100 for $319.00
Add to Cart
09-924-576
View Documents
Cytiva
7182001
0.2 μm 13 mm
Pack of 100 for $226.00
Add to Cart
09-924-575
View Documents
Cytiva
7181002
0.1 μm 25 mm
Pack of 100 for $613.00
Add to Cart
09-905-6
View Documents
Cytiva
7182004
0.2 μm 47 mm
Pack of 100 for $468.00
Add to Cart
09-905-14
View Documents
Cytiva
7184001
0.45 μm 13 mm
Pack of 100 for $237.00
Add to Cart
09-905-55
View Documents
Cytiva
7195002
5 μm 25 mm
Pack of 100 for $317.00
Add to Cart
09-905-19
View Documents
Cytiva
7184009
0.45 μm 90 mm
Pack of 25 for $295.00
Add to Cart
09-924-578
View Documents
Cytiva
7182014
0.2 μm 142 mm
Pack of 25 for $432.00
Add to Cart
09-924-581
View Documents
Cytiva
7188009
0.8 μm 90 mm
Pack of 25 for $303.00
Add to Cart
09-905-5
View Documents
Cytiva
7182002
0.2 μm 25 mm
Pack of 100 for $324.00
Add to Cart
09-924-577
View Documents
Cytiva
7182009
0.2 μm 90 mm
Pack of 25 for $295.00
Add to Cart
09-927-885
View Documents
Cytiva
7184005
0.45 μm 50 mm
Pack of 100 for $381.00
Add to Cart
Description
Description
Recommended for majority of routine applications, this membrane is manufactured under strictly controlled conditions. Whatman™ Type WCN Cellulose Nitrate Membranes narrow pore size distribution for improved surface capture and analysis.
  • Higher strength and flexibility - Most membranes are inherently brittle and difficult to handle; it is not uncommon for filters to be damaged during loading into holders or while in use. Whatman cellulose nitrate membrane filters have a noticeably improved flexibility and are made to tolerate abuse during handling, loading, and autoclaving without sacrificing integrity. These membranes are among the strongest of their type available, as measured and compared by burst pressure tests.
  • Low extractable levels - The level of extractables in membrane filters has become more important with advances in filtration or adsorption techniques. In particular, pharmaceutical, immunological, and biomedical tissue culture and trace analysis applications can be adversely affected by high extractable levels. Whatman cellulose nitrate membrane filters have a low level of extractables, generally below that of other membranes of a similar type.
  • Narrow pore size distribution - One of the major features of Whatman membrane filters is the narrow distribution of pore sizes. The rated pore size of these membranes is closely controlled due to the advanced manufacturing and control system. Additionally, the batch-to-batch variation is minimized providing more consistent laboratory results.
  • Increased temperature stability - Membrane filters are normally autoclaved at 121°C without loss of integrity. Cellulose nitrate membranes are supplied as circles, sheets, or reels.
  • Reduced shrinkage - Excessive shrinkage can cause problems during autoclaving and is often the cause of membranes tearing in their holders after autoclaving. It may also cause a reduction in flow rate and total throughput. Whatman membranes exhibit a low shrinkage during autoclaving.
  • White plain filters - This is the standard membrane filter for the majority of laboratory applications involving particles and cells in the range of 0.1 μm to 12.0 μm. The residue after filtration is found to be almost completely on the surface of the membrane and allows physical recovery of deposits and microscopic examination.
Specifications
Specifications
Autoclavable
White
Cellulose Nitrate
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.