Learn More
Qiagen Uni-Core Punch
Disposable punch for use with FTA cards, FTA Elute Cards, and EasiCollect device
$82.00 - $468.00
Specifications
For Use With (Application) Whatman™ FTA™ cards, FTA™ elute cards and EasiCollect device
Includes Two cutting mats
Product Type Accessory, FTA Card
View More Specs
Products 3
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
05-715-109
View Documents
Qiagen
WB100039
3.0mm
Pack of 4 for $468.00
Add to cart
05-715-110
View Documents
Qiagen
WB100040
6.0mm
Pack of 4 for $82.00
Add to cart
09-924-589
View Documents
Qiagen
WB100029
2.0mm
Pack of 4 for $86.00
Add to cart
Description
Description
Disposable punch for use with FTA cards, FTA Elute Cards, and EasiCollect device
  • Punch with polished steel tip that is case-hardened and can be sterilized
  • Stainless steel cutting edges enable manual coring, retrieval, and controlled ejection of FTA punch
  • Housed in plastic handle
  • Each tip provides up to 500 punches before disposal
  • Available in multiple diameters varying from 0.35 to 12mm
  • Manual punching with low sample volumes punching 1 to 50 samples per day
  • Minimum contamination and carryover between samples when cleaning procedure is followed
Specifications
Specifications
Whatman™ FTA™ cards, FTA™ elute cards and EasiCollect device
Two cutting mats
Accessory, FTA Card