Learn More
Cytiva Whatman™ UniFilter™ 96-Well, 800μL Microplates
Catalog No. PF77002810 Shop All Cytiva Products
Click to view available options
Membrane:
Polypropylene 0.45μm
Whatman GF/B
Whatman GF/F
Suited for purifications, isolations, and separation of biomolecules, such as DNA
  • Clear polystyrene plates available with a variety of filter media
  • Integral filter design prevents well-to-well crosstalk
  • Unique drip director design on the bottom of the plates controls filtrate placement
    • Long drip directors eliminate well-to-well contamination during the filtration process
    • Short drip directors are effective for screening or filtering to waste
  • Standard footprint is compatible with most plate readers, autosamplers, and robots
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Membrane Whatman GF/F
Description UNIFILTER 96-well microplate, 800 μl, long drip director
Array 8 x 12
Color Clear
Material Polystyrene
No. of Wells 96
Well Shape Filter
For Use With (Application) Can be used with both vacuum and centrifuge techniques, making it a vital and flexible tool in every high-throughput laboratory
Includes Long drip director
Quantity 25/Pk.
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.