Learn More
Cytiva Whatman™ UniFilter™ 96-Well, 800μL Microplates
Suited for purifications, isolations, and separation of biomolecules, such as DNA
$1576.00 - $1689.00
Specifications
Format | Standard |
---|---|
Description | UNIFILTER 96-well microplate, 800 μl, long drip director |
Array | 8 x 12 |
Color | Clear |
Material | Polystyrene |
Catalog Number | Mfr. No. | Membrane | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Membrane | Price | Quantity & Availability | |||||
PF77002803
|
Cytiva
77002803 |
Whatman GF/B |
Pack of 25 for $1,689.00
|
|
|||||
PF77002810
|
Cytiva
77002810 |
Whatman GF/F |
Pack of 25 for $1,620.00
|
|
|||||
PF77002805
|
Cytiva
77002805 |
Polypropylene 0.45μm |
Pack of 25 for $1,576.00
|
|
|||||
Description
Suited for purifications, isolations, and separation of biomolecules, such as DNA
- Clear polystyrene plates available with a variety of filter media
- Integral filter design prevents well-to-well crosstalk
- Unique drip director design on the bottom of the plates controls filtrate placement
- Long drip directors eliminate well-to-well contamination during the filtration process
- Short drip directors are effective for screening or filtering to waste
- Standard footprint is compatible with most plate readers, autosamplers, and robots
Specifications
Standard | |
8 x 12 | |
Polystyrene | |
Filter | |
25/Pk. |
UNIFILTER 96-well microplate, 800 μl, long drip director | |
Clear | |
96 | |
Long drip director |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.