Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Whatman™ UniFilter™ Microplates, 24-Well, 10mL

Ideal for applications that require larger sample or reagent volumes (e.g. biomolecular purification by solid-phase extraction and organic synthesis

Manufacturer:  GE Healthcare 77009904

 View more versions of this product

Catalog No. 05-713-849


Add to cart

Description

Description

  • Natural polypropylene plates
  • Patented integral filter design prevents well-to-well cross contamination
  • Standard footprint—compatible with most plate readers, autosamplers and robots
  • Unique drip director design on the bottom of the plates controls filtrate placement
  • Available in several media types

Specifications

Specifications

Filter
The 10 mL UNIFILTER is used with large sample or reagent volumes. Typical applications include biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation.
Long drip director and MBPP Filter
25-30 μm
UNIFILTER Microplate, 24-well, 10 mL, 25 to 30 μm melt blown polypropylene, natural polypropylene, filter bottom with long drip director
10000 μL
24
Clear
Biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation
Polypropylene
25/Pk.
Non-sterile
Standard
4 x 6
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit