Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ UniFilter™ Microplates, 24-Well, 10mL

Ideal for applications that require larger sample or reagent volumes (e.g. biomolecular purification by solid-phase extraction and organic synthesis

Supplier:  Cytiva 77009917

 View more versions of this product

Catalog No. 09-927-350


Explore available promotions
Add to Cart

Description

Description

  • Natural polypropylene plates
  • Patented integral filter design prevents well-to-well cross contamination
  • Standard footprint—compatible with most plate readers, autosamplers and robots
  • Unique drip director design on the bottom of the plates controls filtrate placement
  • Available in several media types

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
4 x 6
Natural
Polypropylene
24
Biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation
The 10 mL UNIFILTER is used with large sample or reagent volumes. Typical applications include biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation.
10 to 12 μm
UNIFILTER Microplate, 24-well, 10 mL, 10 to 12 μm melt blown polypropylene, natural polypropylene
10,000 μL
Filter
Long drip director and MBPP Filter
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.