Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ UNIPLATE™ Microplates

Manufactured from polypropylene for superior chemical and thermal stability. Whatman™ UNIPLATE™ Microplates are suitable for simple filtrate collection and homogeneous assay techniques used in high-throughput screening (HTS).

$550.00 - $643.00

Specifications

Format Standard
Description UNIPLATE microplates
Color Natural
Material Polypropylene
For Use With (Application) Ideal for homogeneous assay techniques utilized in high-throughput screening
View More Specs

Products 3
Catalog Number Mfr. No. Volume (Metric) Well Well Shape No. of Wells Sterility Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Well Well Shape No. of Wells Sterility Price Quantity & Availability  
PF77015102
View Documents
Cytiva
77015102
10 mL Round 24 Non-sterile
Pack of 25 for $643.00
Edge
Add to Cart
 
PF77015500
View Documents
Cytiva
77015500
5 mL Flat Bottom (F-well) 48 Non-sterile
Pack of 25 for $551.00
Add to Cart
 
PF77015110
View Documents
Cytiva
77015110
10 mL Round 24 Sterile
Pack of 25 for $550.00
Add to Cart
 
Description

Description

Manufactured from polypropylene for superior chemical and thermal stability. Whatman™ UNIPLATE™ Microplates are suitable for simple filtrate collection and homogeneous assay techniques used in high-throughput screening (HTS).

  • Withstand low temperatures (-80°C) for long-term storage
  • Conform to ANSI™/SBS standards for compatibility with most microplate readers and automated plate-handling devices
Specifications

Specifications

Standard
Natural
Ideal for homogeneous assay techniques utilized in high-throughput screening
UNIPLATE microplates
Polypropylene
25/Pk.
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit