Learn More
GE Healthcare Whatman™ UniPrep™ Syringeless Filters - PVDF
Process samples in one-third the time of conventional syringe filters
Manufacturer:  GE Healthcare UN113UAQU
 View more versions of this product
Catalog No. 09-923-1
Add to cart
Description
Description
UniPrep syringeless filters are preassembled filtration devices for the filtration and storage of laboratory samples.
  • PVDF filter: low protein binding membrane for filtration of samples with aqueous or aqueous/organic solvent composition
  • Consists of external sample test tube, plunger (which incorporates membrane filter and prefilter), removable cap
  • Filtration area: 0.3cm2
  • Safe, efficient—prefilter and membrane can't pop off
  • Air purge unnecessary—inserting plunger into tube provides a positive pressure purge of sample through filter
  • Sealed glass microfiber prefilter traps larger particles before they can clog the membrane
Applications:
  • Increase lab productivity and efficiency
  • Generate less waste
  • Realize greater overall cost savings per test
Specifications
Specifications
Syringeless Filter
PVDF
0.45μm
17mm
50μL
0.28cm sq.