Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
GE Healthcare Whatman™ UniSeal™ Flexible Capmat
Manufacturer:  GE Healthcare 77040004
 View more versions of this product
Catalog No. 09-003-42
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
·    Positively seals top of each well individually; prevents crosstalk and minimizes evaporation
·    May be used on filter or collection microplates
·    Autoclavable to 120°C (248°F); can be stored in temperatures from -40° to +95°C (-40° to +203°F)
·    Pierceable