Learn More
Whatman™ ZC Syringe Filters
Glass fiber prefilters preclean samples for greater throughput
Manufacturer:  Whatman™ 68442502
 View more versions of this product
Catalog No. 09-927-4A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Syringe Filter
Polytetrafluoroethylene (PTFE)
>10mL
25mm
Can be used in BenchMate and other Zymark robotic systems
0.2μm