Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ 1000mL Media Bottle, Clear Type I
SDP

Catalog No. 02912652 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Liner:
Polyethylene
Rubber
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Liner
02-912-652 Rubber
02-912-650 Polyethylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-912-652 Supplier DWK Life Sciences Supplier No. W219580
Add to Cart
Add to Cart

Manufactured from borosilicate glass which provides the best chemical resistance. Wheaton 1000mL Media Bottle, Clear Type I are available with black phenolic Polyethylene or Rubber lined caps.

  • Clear borosilicate glass bottles, non-graduated
  • Special pour lip facilitates pouring
  • Packaged in partitioned carton to help prevent scratch marks on bottles
  • Hazardous communication label included for each case for content ID and safety notification
  • Tamper evident seal keeps lidded bottles sealed to protect contents from accidental misuse
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
Color Clear
Includes Pour lip
Material Borosilicate Glass (Type I)
Packaging Partitioned case
Quantity 12 EA
Closure Color Black
Closure Included Yes
Height (Metric) 231 mm
Product Type Media Bottle
Closure Liner Rubber
Closure Material Phenolic
Closure Size 38-430 mm
Diameter (Metric) 103 mm
Closure Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.