Learn More
Wheaton™ 12-Channel Modules for Acura Electro™ 956 Pipetters
For Acura Electro 956 12-Channel Pipetters
Manufacturer:  Wheaton™ W870142
 View more versions of this product
Catalog No. 03-410-194
  Check Availability
Add to cart
Specifications
Description
  • One control unit fits a choice of 27 volumetric assemblies available separately greatly extends working possibilities and makes electronic pipetting affordable to all budgets
  • Disassembling and re-assembling without any tool contributes towards maximal flexibility
  • Calibration specificities of extra modules, as set by factory QC or by pipette owner, are retained in instrument memory.
Description & Specifications
Specifications
±3.5%
Universal fit pipette tip
0.05μL
2 Years
12
0.5 to 10μL
Rechargeable
Ergonomic
10μL
Acura™ Electro™ 956
12-Channel Module for Pipetter
0.5 to 10μL