Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ 12 x 32mm Polypropylene Vials with Limited Volume Insert Available on GSA/VA Contract for Federal Government customers only.

Limited Volume

Manufacturer:  DWK Life Sciences 225180

 View more versions of this product

Catalog No. 03-336-1


Add to cart

Description

Description

  • 0.30mL volume
  • Offer good chemical resistance
  • Conical interior eliminates time-consuming assembly of vials and limited volume inserts
  • Designed for maximum retrieval of contents

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

11mm
For maximum retrieval of contents
32mm
1000 Case
12 mm
0.3 mL
Crimp
1.25 in.
Limited volume insert
Autosampler Vial
Polypropylene
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit