Wheaton™ 25mm Filtration Assemblies and Accessories Available on GSA/VA Contract for Federal Government customers only.
Choose from a wide range of filtration assemblies and accessories to suit your application needs
Manufacturer: Wheaton™ 419332
 View more versions of this product
Catalog No. 14-209-101
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance USP Type I, ASTM E 438 Type I, Class A
    Material Glass
    Product Type Fritted Glass Base
    For Use With (Equipment) 25mm filtration assembly
    Pore Size (Metric) 40 to 60μm
    Quantity (No. per Case) 1 Case
  • Description & Specifications
Related Products
Product Content Feedback