Learn More
DWK Life Sciences Wheaton™ ABC Amber Glass Vial Convenience Packs
40% larger opening improves sample accessibility while reducing autosampler needle damage
$92.30 - $135.90
Specifications
Diameter (English) Outer | 0.47 in. |
---|---|
Diameter (Metric) Outer | 12 mm |
Material | Borosilicate Glass |
Volume (Metric) | 1.8 mL |
Height (English) | 1.25 in. |
Catalog Number | Mfr. No. | Closure Color | Septum | Marking Spot | Pre-slit | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Closure Color | Septum | Marking Spot | Pre-slit | Price | Quantity & Availability | ||
02-911-663
|
DWK Life Sciences
W22515704 |
Natural | Blue PTFE, White Silicone | Yes | No |
Case of 100 for $120.30
|
|
||
02-911-661
|
DWK Life Sciences
W22515604 |
Natural | Blue PTFE, White Silicone | No | No |
Case of 100 for $118.30
|
|
||
02-911-662
|
DWK Life Sciences
W2251560401 |
Black | Blue PTFE, White Silicone | No | No |
Case of 100 for $118.30
|
|
||
02-911-664
|
DWK Life Sciences
W2251570401 |
Black | Blue PTFE, White Silicone | Yes | No |
Case of 100 for $120.30
|
|
||
02-912-920
|
DWK Life Sciences
W2251560101 |
Black | White PTFE, Red Silicone | No | No |
Case of 100 for $92.30
|
|
||
02-912-921
|
DWK Life Sciences
W2251560103 |
Red | White PTFE, Red Silicone | No | No |
Case of 100 for $92.30
|
|
||
02-912-922
|
DWK Life Sciences
W2251560104 |
Blue | White PTFE, Red Silicone | No | No |
Case of 100 for $92.30
|
|
||
02-912-919
|
DWK Life Sciences
W22515601 |
Natural | White PTFE, Red Silicone | No | No |
Case of 100 for $92.30
|
|
||
02-912-923
|
DWK Life Sciences
W2251560105 |
Yellow | White PTFE, Red Silicone | No | No |
Case of 100 for $92.30
|
|
||
02-912-926
|
DWK Life Sciences
W2251570101 |
Black | White PTFE, Red Silicone | Yes | No |
Case of 100 for $97.50
|
|
||
02-912-925
|
DWK Life Sciences
W22515701 |
Natural | White PTFE, Red Silicone | Yes | No |
Case of 100 for $97.50
|
|
||
02-912-924
|
DWK Life Sciences
W2251560107 |
Green | White PTFE, Red Silicone | No | No |
Case of 100 for $92.30
|
|
||
02-912-927
|
DWK Life Sciences
W2251570103 |
Red | White PTFE, Red Silicone | Yes | No |
Case of 100 for $94.00
|
|
||
02-912-929
|
DWK Life Sciences
W2251570105 |
Yellow | White PTFE, Red Silicone | Yes | No |
Case of 100 for $97.50
|
|
||
02-912-928
|
DWK Life Sciences
W2251570104 |
Blue | White PTFE, Red Silicone | Yes | No |
Case of 100 for $97.50
|
|
||
02-912-930
|
DWK Life Sciences
W2251570107 |
Green | White PTFE, Red Silicone | Yes | No |
Case of 100 for $97.50
|
|
||
03-410-736
|
DWK Life Sciences
W2251530804 |
Blue | Red PTFE, White Silicone | Yes | Yes |
Case of 100 for $135.90
|
|
||
03-410-735
|
DWK Life Sciences
W2251520804 |
Blue | Red PTFE, White Silicone | No | Yes |
Case of 100 for $128.60
|
|
||
Description
- Manufactured from amber Type I borosilicate glass
- 12 x 32 mm screw top with 9 mm ABC screw cap
- Available with or without writing patch
Specifications
0.47 in. | |
Borosilicate Glass | |
1.25 in. | |
Screw Cap | |
9 mm |
12 mm | |
1.8 mL | |
Polypropylene | |
32 mm |