Learn More
DWK Life Sciences Wheaton™ ABC Vial Convenience Packs: Amber Glass
40% larger opening improves sample accessibility while reducing autosampler needle damage
Manufacturer:  DWK Life Sciences W2251560107
 View more versions of this product
Catalog No. 02-912-924
Add to cart
Description
Description
  • Manufactured from amber Type I borosilicate glass
  • 12 x 32mm screw top with 9mm ABC screw cap
  • Available with or without writing patch
Specifications
Specifications
Green
9mm
0.47 in.
32mm
Chromatography Vial
12mm
1.8mL
Polypropylene
Screw cap
1.25 in.
PTFE/Red Silicone
Green
Borosilicate Glass