Learn More
DWK Life Sciences Wheaton™ Acura Electro™ 926 XS and 936 Single Channel Pipette - Basic Package
Includes a microprocessor-controlled motor and rechargeable battery pack
Manufacturer:  DWK Life Sciences W870908A
 View more versions of this product
Catalog No. 03-410-004
Add to cart
Includes: Acura electro basic package includes electronic pipette, individual QC certificate and operating instructions, pipette charging stand, fast charger with AC adapter and cord, additional battery pack, and pipette tip samples.
Description
Description
Reduced shaft length and conical end improve driveability
  • Covers the volume range 0.1µL to 10ml
  • Metrological performance and stability
  • Easy access to microtubes and microplates
W870906-A for use with 200µl pipette tips.
Order Info
Additional interchangeable volumetric modules available as accessories.
Specifications
Specifications
2 Years
1
1 to 20μL
NiMH
Digital
200μL Pipette tip
0.1μL
926XS
1 to 20μL
±1.2 to <±0.5%
200μL Pipette tip
Ergonomic
Electronic pipette, individual QC certificate and operating instructions, pipette charging stand, fast charger with AC adapter and cord, microprocessor controlled motor, additional battery pack and pipette tip samples