Wheaton™ Acura™ Electro 926XS Single Channel Micropipet
Unsurpassed metrological performance and stability
Manufacturer: Wheaton™ W870904
 View more versions of this product
Catalog No. 03-410-102
  Check Availability
Add to Cart
    Specifications
    Accuracy ±1.2 to <±0.6%
    Battery Type NiMH
    Compatible Tips Ultra 10μL pipette tip
    Display Digital
    Finger Style Ergonomic
    For Use With (Equipment) Ultra 10μL pipette tip
    Includes Rechargeable battery pack, individual QC certificate, pipette tip samples and operating instructions
    Increments 0.05μL
    Programmability Self Teaching
    Warranty 2 Years
    Model 926XS
    Number of Channels 1,1
    Volume (Metric) Range 0.5 to 10μL
    Volume (Metric) 0.5 to 10μL
    Compatibility Ultra 10μL pipette tip
    Capacity Range 0.5 to 10μL
  • Description & Specifications
Related Products
Product Content Feedback