Learn More
DWK Life Sciences Wheaton™ Acura™ Electro 926XS Single Channel Micropipet
Unsurpassed metrological performance and stability
Manufacturer:  DWK Life Sciences W870906
 View more versions of this product
Catalog No. 03-410-103
  Check Availability
Add to cart
Description
Description
  • Reduced shaft length and conical end improve driveability
  • Easy access to microtubes and microplates
  • Optional longer volumetric modules
  • Simple conversion by user at any time
  • Optimal ergonomics, light weight
  • Easy, intuitive, self-teaching program
  • Large display, reversible for right or left reading
  • Swift-interchange battery pack
  • Fast charge, long working autonomy
  • Adjustable tip ejector fitting most tips
  • Pipetting cycle counter
  • Speed selector allows swift speed change even during pipetting process
  • Easy tip ejection
Certifications
CE Marked
Compliance
  • Conforms to IVD
  • RoHS compliant
Warranty and Services
Two-years
Specifications
Specifications
±1.2 to <±0.6%
200μL Pipette tip
Ergonomic
Rechargeable battery pack, individual QC certificate, pipette tip samples and operating instructions
Self Teaching
926XS
0.5 to 10μL
NiMH
Digital
200μL Pipette tip
0.05μL
2 Years
1
0.5 to 10μL