Learn More
DWK Life Sciences Wheaton™ Acura electro™ 956 12-Channel Pipetters - Basic Package
Large selection of 12-channel pipettes
Manufacturer:  DWK Life Sciences W870932A
 View more versions of this product
Catalog No. 03-410-016
  Check Availability
Add to cart
Includes: Acura electro basic package includes electronic pipette, individual QC certificate and operating instructions, pipette charging stand, fast charger with AC adapter and cord, additional battery pack, and pipette tip samples.
Description
Description
360° rotation of multichannel volumetric module for best working position
  • Microprocessor-controlled motor
  • Rechargeable battery pack
  • Also fits any single channel volumetric module
  • Models cover the volume range 0.5 to 350µL
Specifications
Specifications
±3.5 to <±1%
Universal fit pipette tip
Ergonomic
0.05μL
2 Years
12
0.5 to 10μL
Rechargeable
Digital
Electronic pipette, individual QC certificate and operating instructions, pipette charging stand, fast charger with AC adapter and cord, additional battery pack and pipette tip samples
10μL
Acura™ Electro™ 956
0.5 to 10μL
missing translation for 'documents'