Learn More
Wheaton™ Acura electro™ 956 12-Channel Pipetters - Basic Package
Large selection of 12-channel pipettes
Manufacturer:  Wheaton™ W870938A
 View more versions of this product
Catalog No. 03-410-019
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
±1 to <±0.6%
Universal fit pipette tip
Ergonomic
5μL
2 Years
12
20 to 350μL
Rechargeable
Digital
Electronic pipette, individual QC certificate and operating instructions, pipette charging stand, fast charger with AC adapter and cord, additional battery pack and pipette tip samples
350μL
Acura™ Electro™ 956
20 to 350μL