Learn More
DWK Life Sciences Wheaton™ Acura Electro™ 956 12-Channel Pipetters
Fits any single channel volumetric module
Manufacturer:  DWK Life Sciences W870932
 View more versions of this product
Catalog No. 03-410-116
  Check Availability
Add to cart
Includes:
  • Microprocessor controlled motor and rechargeable battery pack
  • QC certificate and operating instructions
  • Samples of pipet tips
Description
Description
  • With 360 degree rotation of multichannel volumetric module for best working position
  • Electronic, 4.8V
Warranty and Services
Two year
Specifications
Specifications
±3.5 to <±1%
Universal fit pipette tip
Ergonomic
0.05μL
2 Years
12
Rechargeable
Digital
Microprocessor controlled motor, rechargeable battery pack, QC certificate, operating instructions and samples of pipette tip
10μL
Acura™ Electro™ 956
0.5 to 10μL