Learn More
Wheaton™ Acura™ electro 956 Eight-Channel Pipets
Fits any single channel volumetric module
Manufacturer:  Wheaton™ W870924
 View more versions of this product
Catalog No. 03-410-112
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
±3.5 to <±1%
Universal fit pipette tip
Ergonomic
0.05μL
2 Years
8
0.5 to 10μL
Rechargeable
Digital
Microprocessor controlled motor, rechargeable battery pack, QC certificate, operating instructions and samples of pipette tip
10μL
Acura™ Electro™ 956
0.5 to 10μL