Learn More
DWK Life Sciences Acura™ electro 956 Eight-Channel Pipets
Fits any single channel volumetric module
Supplier: DWK Life Sciences W870926
Includes: Microprocessor-controlled motor, rechargeable battery pack, QC certificate, operating instructions and samples of pipet tips
Description
The microprocessor controlled pipette features intuitive software, eliminating any complicated programming steps. There are six working modes that are easy to access and many volumes to select from.
- For volume range 0.5 to 350μL
- Lightweight and perfect hand fitting
- Sequential tip ejection
Specifications
±1 to <±0.8% | |
Universal fit pipette tip | |
8 | |
Pipette | |
Digital | |
Microprocessor controlled motor, rechargeable battery pack, QC certificate, operating instructions and samples of pipette tip | |
2 Years |
Rechargeable | |
Acura™ 956 | |
Acura™ Electro™ | |
2.5 to 50 μL | |
Ergonomic | |
0.25μL |