Learn More
DWK Life Sciences Wheaton™ Acura™ electro 956 Eight-Channel Pipets
Fits any single channel volumetric module
Manufacturer:  DWK Life Sciences W870928
 View more versions of this product
Catalog No. 03-410-114
  Check Availability
Add to cart
Includes: Microprocessor-controlled motor, rechargeable battery pack, QC certificate, operating instructions and samples of pipet tips
Description
Description
The microprocessor controlled pipette features intuitive software, eliminating any complicated programming steps. There are six working modes that are easy to access and many volumes to select from.
  • For volume range 0.5 to 350µL
  • Lightweight and perfect hand fitting
  • Sequential tip ejection
Specifications
Specifications
±0.9 to <±0.6%
Universal fit pipette tip
Ergonomic
1μL
2 Years
8
10 to 200μL
Rechargeable
Digital
Microprocessor controlled motor, rechargeable battery pack, QC certificate, operating instructions and samples of pipette tip
200μL
Acura™ Electro™ 956
10 to 200μL