Learn More
Wheaton™ Center Disc Tear-Out Aluminum Seals, Unlined Available on GSA/VA Contract for Federal Government customers only.
Tear-out seals remove completely allowing content to pour from bottle. Wheaton™ Center Disc Tear-Our Aluminum Seals, Unlined are available in a wide range of sizes and colors.
Manufacturer:  Wheaton™ 22418201
 View more versions of this product
Catalog No. 06-406-14A
  Check Availability
Add to cart
Specifications
Description
  • Good barrier properties
  • Color selection includes natural, red, blue and green
  • Chemically resistant to alcohol and acetone, among others
Description & Specifications
Specifications
Center Disc Tear-out
Natural
Seal
13mm
Aluminum
1000/Cs.
Aluminum