Learn More
Wheaton™ Tear-Off Aluminum Seals, Unlined
Tear-off seals remove completely allowing content to pour from bottle. Wheaton™ Tear-Off Aluminum Seals, Unlined are available in a wide range of sizes and colors.
Manufacturer:  Wheaton™ 22419301
 View more versions of this product
Catalog No. 06-406-15
  Check Availability
Add to cart
Specifications
Description
  • Good barrier properties
  • Color selection includes natural, red, blue and green
  • Chemically resistant to alcohol and acetone, among many others
Description & Specifications
Specifications
Complete Tear-off
Natural
Seal
20mm
Aluminum
1000/Cs.
Aluminum