Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wheaton™ Amber Boston Rounds with PE Cone Lined Caps

Amber Type III soda lime glass protects light-sensitive solutions

Manufacturer:  Wheaton™ W216852

 View more versions of this product

Catalog No. 02-911-366


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

16 oz., 500mL
16 oz.
Polyethylene
2.95 in.
Soda Lime Glass
28-400
168mm
500mL
Phenolic
Amber
6.6 in.
Black
75mm
Screw
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit