Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Amber Boston Rounds with Rubber-lined Caps

Amber glass protects light-sensitive solutions

Manufacturer:  DWK Life Sciences W216855

 View more versions of this product

Catalog No. 02-911-369


  Check Availability
Add to cart

Description

Description

  • Type III soda-lime glass

Specifications

Specifications

4 oz., 125mL
4 oz.
Rubber
1.88 in.
Glass
22-400
112mm
125mL
Phenolic
Amber
4.4 in.
Black
48mm
Screw
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit