Learn More
DWK Life Sciences Wheaton™ Amber Glass E-Z Vials Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  DWK Life Sciences W2251730207
 View more versions of this product
Catalog No. 02-926-955
Add to cart
Description
Description
  • Manufactured from certified clear and amber Type I borosilicate glass
  • Vials are packaged 100 per shelf pack and 10 packs per case
  • 40% larger opening improves sample accessibility while reducing autosampler needle damage
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Yes
0.005 PTFE/ 0.05 silicone r/w liner
Green
Borosilicate Glass