Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Amber Wide-Mouth Packers with Poly-Seal™-lined Closures

Amber glass for storing light-sensitive powders

Manufacturer:  DWK Life Sciences W216951

 View more versions of this product

Catalog No. 02-912-148


Add to cart

Description

Description

  • USP Type III soda-lime glass
  • Allows for easy removal of contents

Specifications

Specifications

1 oz., 30mL
1 oz.
Polyseal
1.45 in.
Soda Lime Glass
28-400
67mm
30mL
Phenolic
Amber
2.63 in.
Black
37mm
Screw
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit