Learn More
Wheaton™ Black Phenolic Screw Caps with PTFE faced 14B Rubber Liners for E-Z Ex-Traction™ Vials Available on GSA/VA Contract for Federal Government customers only.
Designed to ensure the ultimate in product safety
Manufacturer:  Wheaton™ 240418
 View more versions of this product
Catalog No. 06-406-35
  Check Availability
Add to cart
Specifications
Description
  • Autoclavable
  • Liner made of styrene-butadiene rubber faced with PTFE
  • Excellent chemical resistance
Description & Specifications
Specifications
ISO 9001
Black
20mL Vial
PTFE/Rubber
100 Cs.
24-400