Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton™ Blue Starline Polypropylene Caps for Narrow-Mouth Containers
Click to view available options
Neck Finish:
20-410
24-410
28-410
38-430
No. per Case:
48 Cs.
72 Cs.
Description
- Leak-proof design
- Autoclavable
Specifications
For Use With (Equipment) | Redi-pak™ leak resistant narrow mouth bottle |
Color | Blue |
Material | Polypropylene |
No. per Case | 72 Cs. |
Neck Finish | 20-410 |
Certifications/Compliance | ISO 9001 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction