Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Wheaton™ BOD Bottles
Available with either glass robotic or pennyhead stoppers
Manufacturer:  Wheaton™ 22749799G
 View more versions of this product
Catalog No. 02-911-305
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Yes
10.14 oz.
6.9cm
16.5cm
Used to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action
Yes
10.14 oz., 300mL
300mL
2.7 in.
6.49 in.
Borosilicate Glass
Specials
Pennyhead