Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Bottles Amber Wide-Mouth Packers with Polyvinyl-lined Closures

Amber glass for storing light-sensitive powders. Wheaton™ Bottles Amber Wide-Mouth Packers with Polyvinyl-lined Closures are available in several capacities.

Manufacturer:  DWK Life Sciences W216958

 View more versions of this product

Catalog No. 02-912-204


  Check Availability
Add to cart

Description

Description

  • USP Type III soda-lime glass
  • Allows for easy removal of contents

Specifications

Specifications

16 oz., 500mL
16 oz.
Poly/Vinyl
3.14 in.
60/Cs.
53-400
146mm
500mL
Polypropylene
Amber
5.74 in.
White
80mm
Screw
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit