Learn More
DWK Life Sciences Wheaton™ Bottom-Drive R2P 2.0 Roller Apparatus
Advanced Production Scale Roller Culture Equipment
Manufacturer:  DWK Life Sciences WRBPR5010A
 View more versions of this product
Catalog No. 02-924-936
  Check Availability
Add to cart
Description
Description
  • Color touch screen or digital display
  • Icon driven interface with intuitive menus
  • Surface mount gold plated electrical connections for better reliability in high temperature environments
  • Updated industry standard networking communications protocols and connections
  • Lower power consumption for longer battery run time and safer design
  • Backwards compatibility for continuing support of existing installations
  • Retrofit kits available to update older Wheaton systems or unsupported systems
  • Maintenance free brushless DC motors for years of trouble-free operation
Specifications
Specifications
W x H: 79 x 35cm (31 x 14 in.)
63cm
Bottom Drive Roller Culture System with 1 deck
5 Bottle positions
For laboratory, pilot plant and production cell culture installations
6 in.
6 in.
25 in.
120VAC
R2P 2.0
1