Learn More
DWK Life Sciences Wheaton™ Bottom-Drive Standard Roller Apparatus
For anchorage dependent cell growth in roller bottles
Manufacturer:  DWK Life Sciences WSBMR5081A
 View more versions of this product
Catalog No. 02-924-917
  Check Availability
Add to cart
Description
Description
  • Digital display
  • Surface mount gold plated electrical connections
  • Lower power consumption for longer battery run time
  • Backwards compatibility for continuing support
For laboratory, pilot plant and production cell culture installations.
Specifications
Specifications
7 in.
For laboratory, pilot plant and production cell culture installations
7 in.
40 Bottle positions, Options
8