Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wheaton™ Calibrex™ Orango 525 Bottle-Top Dispensers

Manufacturer:  Wheaton™ W844092

 View more versions of this product

Catalog No. 13-709-521


  Check Availability
Add to cart

Specifications

Description

  • Permanent fluid path visibility
  • Integrated calibration system
  • Choice of adjustment button with each unit
  • Long lasting performance stability
  • Autoclavable at 121°C / 250°F fully assembled
  • Two year warranty
Description & Specifications

Specifications

5 to 50mL
<±1.5 to <±0.6%
5 to 50mL
Bottletop Dispenser
Orango 525
2 Years
Yes
<0.5 to <0.1%
Yellow
45mm base thread, 120mm FEP/PCTFE delivery jet and ETFE stopper, 350mm FEP feed tube, alternative screw type volume setting button, 32, 38 and 40mm bottle neck adapters, QC certificate, operating instructions
1mL
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit