Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wheaton™ Calibrex™ Orango 525 Bottle-Top Dispensers w/Flow Control Stopcock

Manufacturer:  Wheaton™ W844100

 View more versions of this product

Catalog No. 13-709-537


  Check Availability
Add to cart

Specifications

Description

  • With Flow Control Stopcock
  • Permanent fluid path visibility
  • Integrated calibration system
  • Choice of adjustment button with each unit
  • Long lasting performance stability
  • Autoclavable at 121°C / 250°F fully assembled
  • Two year warranty
Description & Specifications

Specifications

Yes
Yellow
Ground glass plunger
10 to 100mL
Bottle Top Dispenser
525 Organo
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit