Learn More
DWK Life Sciences Wheaton Caps For Liquid Scintillation Vials
$210.00 - $505.00
Specifications
Closure Type | Screw Cap |
---|---|
For Use With (Equipment) | Liquid Scintillation Vial |
Catalog Number | Mfr. No. | Closure Material | Closure Size | Liner Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Closure Material | Closure Size | Liner Material | Price | Quantity & Availability | |||
06-450-88
![]() |
DWK Life Sciences
240817 |
Urea | 22-400 | Polyethylene Disc |
Case of 1000 for $411.00
|
|
06-450-91
|
DWK Life Sciences
241009 |
Urea | 15-425 | Metal Foil |
Case of 1000 for $213.00
|
|
06-450-90
![]() |
DWK Life Sciences
240917 |
Urea | 22-400 | Polyseal™ |
Case of 1000 for $505.00
|
|
06-450-92
|
DWK Life Sciences
241017 |
Urea | 22-400 | Metal Foil |
Case of 1000 for $351.00
|
|
06-450-94
|
DWK Life Sciences
241317 |
Polypropylene | 22-400 | Linerless |
Case for $210.00
|
|
06-450-86
![]() |
DWK Life Sciences
240804 |
Polypropylene | 22-400 | Metal Foil |
Case of 1000 for $226.00
|
|
Description
- Screw caps for WHEATON liquid scintillation vials
- Case of 1000 provides an economical alterative
- Not autoclavable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Screw Cap | |
Liquid Scintillation Vial |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.