Learn More
DWK Life Sciences Wheaton™ Celstir™ Spinner Double Side Arm Flasks without Water Jacket
Provide maximum surface between culture and glass flask
Supplier: DWK Life Sciences 356884
Description
Provide maximum surface between culture and glass flask
- PTFE-resin coated glass impeller does not protrude through cap and has bottom dimple for more efficient stirring
- Silicone rubber-lined cap
- Autoclavable
Specifications
Silicone Rubber | |
1000 mL | |
PTFE Resin | |
10 to 44 cm |
4.5 cm | |
Flask, impeller and caps | |
Autoclavable |