Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton™ Clear Boston Rounds with PE Cone-Lined Caps
Click to view available options
Capacity:
1 oz., 30mL
16 oz., 500 mL
2 oz., 60 mL
32 oz., 1000 mL
4 oz., 125 mL
8 oz., 250 mL
Diameter (English):
1.22 in.
1.53 in.
1.88 in.
2.36 in.
2.95 in.
3.7 in.
Height (English):
3.1 in.
3.7 in.
4.4 in.
5.43 in.
6.6 in.
8.18 in.
Description
- Well suited for storage and sampling of liquids; clear Type III soda-lime glass provides visibility of contents
- Narrow mouth enables liquids to be carefully poured to prevent spillage
Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage.
Order Info
Caps can be ordered separately.
Specifications
Closure Type | Screw Cap |
Closure Liner | Polyethylene |
Closure Material | Phenolic |
Closure Size | 20-400 mm |
Diameter (Metric) | 31 mm |
Height (Metric) | 79 mm |
Height (English) | 3.1 in. |
Capacity | 1 oz., 30mL |
Capacity (Metric) | 30 mL |
Capacity (English) | 1 oz. |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction