Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Clear Straight-Sided Jars with Polyvinyl-lined Caps

Ideal for soil sampling and environmental applications

Manufacturer:  DWK Life Sciences W216918

 View more versions of this product

Catalog No. 02-911-421

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass

  • Wide mouths provide easy access to contents when using spoons or spatulas

Each case of convenience pack bottles includes hazard communication labels for proper content identification and tamper evident labels to protect sample integrity in transit or long-term storage

Order Info

Caps can be ordered separately.

Compliance

Clear-USP Type III soda-lime glass

Specifications

Specifications

32 oz., 1000mL
32 oz.
Poly/Vinyl
3.74 in.
Soda Lime Glass
White
95mm
Screw
1000mL
Polypropylene
Clear
6.69 in.
Valu-bulk pack
89-400
170mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit