Learn More
DWK Life Sciences WHEATON CompletePAK Clear Kits
The WHEATON CompletePAK product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
$1114.00 - $1365.00
Specifications
Color | Clear |
---|---|
Product Line | Wheaton™ CompletePAK |
Sterility | Sterile |
Includes | Vial, Stopper, and Seal |
Catalog Number | Mfr. No. | Volume (Metric) | Closure Type | Color (Seal) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Closure Type | Color (Seal) | Price | Quantity & Availability | |||
06-451-220
![]() |
DWK Life Sciences
W990000CP |
2 mL | Ultra Pure Sterile | Blue |
Case of 210 for $1,114.00
|
|
06-451-221
|
DWK Life Sciences
W990001CP |
2 mL | Omni Flex 3G | Blue |
Case of 210 for $1,220.00
|
|
06-451-222
|
DWK Life Sciences
W990002CP |
2 mL | OmniFlex 3G Sterile | Blue |
Case of 210 for $1,253.00
|
|
06-451-223
|
DWK Life Sciences
W990003CP |
2 mL | Ultra Pure Sterile | Red |
Case of 210 for $1,114.00
|
|
06-451-224
|
DWK Life Sciences
W990004CP |
2 mL | Omni Flex 3G | Red |
Case of 210 for $1,220.00
|
|
06-451-225
|
DWK Life Sciences
W990005CP |
2 mL | OmniFlex 3G Sterile | Red |
Case of 210 for $1,253.00
|
|
06-451-226
![]() |
DWK Life Sciences
W990006CP |
5 mL | Ultra Pure Sterile Serum Stopper | Blue |
Case of 200 for $1,170.00
|
|
06-451-227
|
DWK Life Sciences
W990007CP |
5 mL | OmniFlex 3G Sterile Serum Stopper | Blue |
Case of 200 for $1,275.00
|
|
06-451-228
|
DWK Life Sciences
W990008CP |
5 mL | OmniFlex 3G Igloo Lyo Stopper | Blue |
Case of 200 for $1,309.00
|
|
06-451-229
|
DWK Life Sciences
W990009CP |
5 mL | Ultra Pure Sterile Serum Stopper | Red |
Case of 200 for $1,170.00
|
|
06-451-230
|
DWK Life Sciences
W990010CP |
5 mL | OmniFlex 3G Sterile Serum Stopper | Red |
Case of 200 for $1,275.00
|
|
06-451-231
|
DWK Life Sciences
W990011CP |
5 mL | OmniFlex 3G Sterile Igloo Lyo Stopper | Red |
Case of 200 for $1,309.00
|
|
06-451-232
|
DWK Life Sciences
W990012CP |
10 mL | Ultra Pure Sterile Serum Stopper | Blue |
Case of 158 for $1,225.00
|
|
06-451-233
|
DWK Life Sciences
W990013CP |
10 mL | OmniFlex 3G Sterile Serum Stopper | Blue |
Case of 158 for $1,331.00
|
|
06-451-234
|
DWK Life Sciences
W990014CP |
10 mL | OmniFlex 3G Sterile Igloo Lyo Stopper | Blue |
Case of 158 for $1,365.00
|
|
06-451-235
|
DWK Life Sciences
W990015CP |
10 mL | Ultra Pure Sterile Serum Stopper | Red |
Case of 158 for $1,225.00
|
|
06-451-236
|
DWK Life Sciences
W990016CP |
10 mL | OmniFlex 3G Sterile Serum Stopper | Red |
Case of 158 for $1,331.00
|
|
06-451-237
|
DWK Life Sciences
W990017CP |
10 mL | OmniFlex 3G Sterile Igloo Lyo Stopper | Red |
Case of 158 for $1,365.00
|
|
Description
- Sterilized, off-the-shelf, ready-to-use (RtU) products
- Variety of Kitted and stand-alone sterile components a solution for any application
- Kitted solutions feature equal amounts of RtU components for reduced scrap rates
- Complete manufacturing & certificate of analysis traceability
- USP 788/EP 2.9.19 Particulate Matter in Injections, USP 85/EP 2.6.1, The Bacterial Endotoxins Test and USP 71/ EP 2.6.14 The Sterility Test, USP 660/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
- CompletePAK ModPAK Bulk configurations
- Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
- Container Closure Integrity (CCI) certification available upon request
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Clear | |
Wheaton™ CompletePAK | |
Sterile | |
Vial, Stopper, and Seal |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.