Wheaton™ Crimpenstein™ Electric Crimper/Decapper - Non-Standard Voltage
Lightweight and easy to operate
Manufacturer: Wheaton™ W225643-D
 View all options for this product
Catalog No. 02-912-850
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    For Use With 13mm Flip-cap Aluminum Seals
    Type Crimpenstein with 13mm Crimper and Decapper Jaw-Set; U.K. plug
    For Use With (Application) Attaching and removing aluminum seals on crimp top/serum finish bottles and vials
    Wheaton Crimpenstein Electric Crimper/Decapper
    • Easily attaches and removes aluminum seals on crimp top/serum finish bottles and vials to improve productivity
    • Adjustable jaw intensity with memory allows user to regulate crimper and decapper jaw action
    • Quick-change jaw-sets available in 11, 13 and 20mm sizes allow for crimping and decapping of different size bottles and vials
    • Lightweight and easy to operate enables semiautomatic operation for extended periods with less effort
    • Electric powered for quick setup and high volume operation
    For information on using the Crimpenstein with flip cap aluminum seals, visit www.wheatonsci.com/crimp. For information on Wheaton crimp top/serum finish bottles and vials, aluminum seals and rubber stoppers, visit www.fishersci.com.
    Optional sturdy stand (Part No. W225630) holds the Crimpenstein in a fixed vertical position for operations where vials need to be handled one at a time.
  • Description & Specifications
Related Products
Product Content Feedback