Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Crimpenstein™ Electric Crimper/Decapper - Non-Standard Voltage
SDP

Catalog No. 02912836 Shop All DWK Life Sciences Products
Change view
Click to view available options
Product Type:
13 mm Crimper Jaw
13 mm Crimper/Decapper Set
20 mm Crimper Jaw
20 mm Crimper/Decapper Set
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
02-912-836 13 mm Crimper Jaw
02-912-831 20 mm Crimper Jaw
02-912-830 13 mm Crimper Jaw
02-912-832 13 mm Crimper Jaw
02-912-839 20 mm Crimper Jaw
02-912-835 20 mm Crimper Jaw
02-912-833 20 mm Crimper Jaw
02-912-837 20 mm Crimper Jaw
02-912-838 13 mm Crimper Jaw
02-912-834 13 mm Crimper Jaw
02-912-848 13 mm Crimper/Decapper Set
02-912-849 20 mm Crimper/Decapper Set
02-912-851 20 mm Crimper/Decapper Set
02-912-852 13 mm Crimper/Decapper Set
02-912-853 20 mm Crimper/Decapper Set
02-912-854 13 mm Crimper/Decapper Set
02-912-855 20 mm Crimper/Decapper Set
02-912-856 13 mm Crimper/Decapper Set
02-912-857 20 mm Crimper/Decapper Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 02-912-836 Supplier DWK Life Sciences Supplier No. W225637G
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Includes

Each Crimpenstein comes with a detachable power cord, a convenient carrying/storage case, a crimper and deapper jaw set or an individual crimper or decapper jaw.

Lightweight and easy to operate

Wheaton Crimpenstein Electric Crimper/Decapper

  • Easily attaches and removes aluminum seals on crimp top/serum finish bottles and vials to improve productivity
  • Adjustable jaw intensity with memory allows user to regulate crimper and decapper jaw action
  • Quick-change jaw-sets available in 11, 13 and 20mm sizes allow for crimping and decapping of different size bottles and vials
  • Lightweight and easy to operate enables semiautomatic operation for extended periods with less effort
  • Electric powered for quick setup and high volume operation
For information on using the Crimpenstein with flip cap aluminum seals, visit www.wheatonsci.com/crimp. For information on Wheaton crimp top/serum finish bottles and vials, aluminum seals and rubber stoppers, visit www.fishersci.com.

Optional sturdy stand (Part No. W225630) holds the Crimpenstein in a fixed vertical position for operations where vials need to be handled one at a time.

Order Info

Crimper and Decapper Jaws (Part No. W225616-W225628) and sturdy stand (Part No. W225630) are sold separately. For standard voltages, see cat. no. 029110001.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Attaching and removing aluminum seals on crimp top/serum finish bottles and vials
Includes Italy, Chile Plug
For Use With (Equipment) 13 mm Flip-cap Aluminum Seals
Product Type 13 mm Crimper Jaw
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.